DIN 53211 Immersion Cup, Industrial Physics
Supplier: GREAT LAKES GAGES, LLC
VF2071 VF2072 VF2073 VF2074 VF2075 VF2077 VF2213 VF2215 VF2216
10025-122EA 560.99 CAD
10025-122 10025-124 10025-126 10025-128 10025-130 10025-132 10025-142 10025-144 10025-146
DIN 53211 Immersion Cup, Industrial Physics
Sample Containers General Purpose Containers
The DIN 53211 Immersion Cups are available in a range of titanium anodized aluminum or stainless steel formats, all with a fixed stainless steel nozzle (inner cavity) and handle
  • Each cup has a long handle, allowing it to be dipped by hand into a liquid container
  • Design of the cup and orifice eliminate hard to clean recesses
  • Cups are manufactured under continuing quality control procedures
  • Each cup features a unique, engraved serial number
Inner dimensions are similar to DIN 53211. Cups are ideal for measuring coatings and other fluids duringMore Product Information