Ford Viscosity Flow Cup, Industrial Physics
Supplier: GREAT LAKES GAGES, LLC
VF2031 VF2032 VF2033
10025-080EA 690.14 CAD
10025-080 10025-082 10025-084
Ford Viscosity Flow Cup, Industrial Physics
Sample Containers General Purpose Containers
Viscosity Flow Cups are used for measuring the consistency of paints, varnishes, and other similar products
  • A relatively deep well surrounding the top of the cup serves to catch any overflow
  • The design of the cup and orifice eliminate hard to clean recesses
  • The outside dimensions have been chosen to support the TQC stands
  • Cups are manufactured under continuing quality control procedures
  • Each cup features a unique, engraved serial number
Cups are available in titanium anodized aluminum or stainless steel formats. All cups feature a fixed stainless steel nozzle (inner cavity). These viscosity cups are suitable for laboratory use with a stand.
Certifications: Compatible with ASTM D 1200.