Ford Immersion Viscosity Cup, Industrial Physics
Supplier: Industrial Physics
VF2087 VF2084 VF2233
10025-076EA 598.88 CAD
10025-076 10025-064 10025-070
Ford Immersion Viscosity Cup, Industrial Physics
Sample Containers General Purpose Containers
The TQC Ford Immersion Cups are available in titanium anodized aluminum or stainless steel
  • Each cup has a long handle, allowing it to be dipped by hand into a liquid container
  • Design of the cup and orifice eliminate hard to clean recesses
  • Cups are manufactured under continuing quality control procedures
  • Each cup features a unique, engraved serial number
Both material formats of these viscosity cups feature a stainless steel inner cavity fixed nozzle and handle. Ideal for measuring coatings and other fluids during application or production. Each viscosity cup comes with a hard plastic storage case, with protective soft material on the inside.
Certifications: Compatible with ASTM D 1200.