Zahn Immersion Viscosity Cup, Industrial Physics
Supplier: Industrial Physics
VF2226 VF2228 VF2230 VF2229 VF2227
10025-058EA 745.13 CAD
10025-058 10025-034 10025-032 10025-062 10025-060
Zahn Immersion Viscosity Cup, Industrial Physics
Sample Containers General Purpose Containers
The TQC Zahn Immersion Cups are stainless steel viscosity cups with fixed stainless steel inner cavity nozzle and handle
  • Each cup has a long handle, allowing it to be dipped by hand into a liquid container
  • Design of the cup and orifice eliminate hard to clean recesses
  • Cups are manufactured under continuing quality control procedures
  • Each cup features a unique, engraved serial number
Ideal for measuring coatings and other fluids during application or production. Each viscosity cup is supplied with a hard plastic storage case, with protective material on the case interior.
Certifications: Complies with ASTM D1084 D4212.