Formalin 10%, Formal-Fixx™, neutral buffered

Supplier: RICHARD-ALLAN SCIENTIFIC CA

Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol

6764254 9990244
CA89369-332EA 242.78 CAD
CA89369-332 CA89369-348
Formalin 10%, Formal-Fixx™, neutral buffered
Formalin

Produce consistent results and extend life of rotary valve in tissue processors with this 10% neutral buffered foramalin.


  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Pre-filled, leak-resistant cups are available


Available premixed, ready-to-use, or in economical concentrate. Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship.

Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR