TBD-2™ Decalcifier, Epredia
Supplier: RICHARD-ALLAN SCIENTIFIC CA
6764003
6764004
CA89369-404CS
519.76
CAD
CA89369-404
CA89369-406
TBD-2™ Decalcifier, Epredia
Stains
Microscopy and staining reagents
Process specimens up to 48 hours past end point undamaged with this formic acid decalcifier.
- Protects cells from actions of acid with added ingredients
- Permits utilization of routine end point tests, such as calcium oxalate