Cytoslide™ Microscope Slides, Epredia
Supplier: Epredia
5991059 5991051 5991059 5991056
89404-830CS 137.34 CAD
89404-840 89404-830 CA89404-840 89404-836
Cytoslide™ Microscope Slides, Epredia
Microscope Slide Boxes
Improve processing and visualization of small-volume specimens with these cytoslides, for use with Shandon™ EZ Single Cytofunnels™ on the Epredia Cytospin™ cytocentrifuge.
  • Uncoated slides available with circle on front or back
  • Available in boxes of 100 slides