EZ Single Cytofunnel™ with White Filter Cards and Caps, Epredia
Supplier: Epredia
A78710003 A78710020
89370-858CS 481.3 CAD
89370-858 89370-866
EZ Single Cytofunnel™ with White Filter Cards and Caps, Epredia
Filters Centrifugal Filters
Optimize cell yield with the Epredia™ EZ Single Cytofunnel™ with white filter cards and caps.
  • Reduces the risk of exposure to pathogenic samples
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Decreases the possibility of cross-contamination
  • Eliminates the need for metal Epredia™ Cytoclips™
A disposable sample chamber with cap and pre-attached white filter card for high absorbency, this product is meant for single use with all Epredia™ Cytospin™ Cytocentrifuge applications.
Produces more cost-effective, thin-layer preparationsMore Product Information