Super Erecta® Shelf Stationary Security Units, Metro
Supplier: Metropolitan Wire
SEC53C SEC53K3 SEC53S SEC55K3 SEC55S SEC56C SEC56K3 SEC56S SEC53C SEC53K3 SEC53S SEC55K3 SEC55S SEC56C SEC56K3 SEC56S
89369-974EA 3269.46 CAD
89369-974 89369-976 89369-978 89369-980 89369-982 89369-984 89369-986 89369-988 CA89369-974 CA89369-976 CA89369-978 CA89369-980 CA89369-982 CA89369-984 CA89369-986 CA89369-988
Super Erecta® Shelf Stationary Security Units, Metro
Furniture Shelves
Protect valuable materials and sensitive items from loss or pilferage.
  • Easily assembled
  • Heavy-gauge open wire construction keeps contents visible at all times, making it easy to check inventory
  • Double door-each door opens 270 degrees and can be secured along the sides of the unit
  • Available in chrome-plated, stainless steel and Metroseal 3 with Microban®
Chrome-plated shelves are ideal for dry environments with minimal exposure to corrosives. Type 304 stainless steel shelves provide maximum protection in corrosive environments. Metroseal 3 Corrosion-resistant shelves are electroplated carbon steel, with a special epoxy coating. Metroseal 3™ shelves includes antimicrobial product protection, which inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation.