HEPA Filter, Labconco®
Supplier: Labconco
3707902 3838502
82018-212EA 1627.4 CAD
82018-212 97006-996 97007-024
HEPA Filter, Labconco®
Filters Gas Filters
Replacement HEPA filter for use with XPert filtered systems and stations, XPert TXE equipment enclosures, FilterMate portable exhausters, Purifier logic biosafety sabinets, Purifier Cell Logic biosafety cabinets and Double HEPA filtration kits.
  • 99.99% efficient on particles 0.3 µm in size
  • Available in 18.0", 24.0", 30.0", and 42.0" widths
Have aluminum frames with either aluminum separators or separatorless pleats.
Ordering information: Note: Units 5' and 6' wide require two filters.