Econo™ Sterile Graves Vaginal Speculum, Sklar
Supplier: Sklar
96-2605 96-2606 96-2607 96-2603 96-2604 96-2602 96-3000 96-3001 96-3002
10184-656CS 1176.58 CAD
10184-656 10184-658 10184-660 10193-316 10193-806 10193-968 75995-432 75995-434 75995-436
Econo™ Sterile Graves Vaginal Speculum, Sklar
Specula
The Econo™ Sterile Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
  • Ready-to-use
  • No need for cleaning and reprocessing
  • Significantly reduces likelihood of cross-contamination
Speculum is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient. They are available in bulk quantities.