AccuGENE® PBS (phostphate buffered saline) Buffers, Lonza
Supplier: LONZA WALKERSVILLE INC. CA
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
- High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
- Efficient - Ready-made solutions eliminate experiment preparation time
- Flexible - Customized solutions are available to meet individual needs
Phosphate buffered saline, or PBS, is isotonic and non-toxic to most cells making it an excellent buffer for biological experiments. The osmolarity and ion concentrations of the solutions match those of the human body, so is often used to maintain cells for manipulation outside of their regular growth environment during experiments. PBS is also used in solutions that disengage attached and clumped cells.
Store all reagents at 18 to 26°C (64 to 79°F).
Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.