Water, AccuGENE® for molecular biology

Supplier: LONZA WALKERSVILLE INC. CA

Synonyms: Hydrogen oxide

51200 51224 51244
CA12001-380EA 117.55 CAD
CA12001-380 CA12001-400 CA12001-420
Water, AccuGENE® for molecular biology
Water

AccuGENE™ Molecular Biology Water is ideal for a wide range of molecular biology applications. 18 megOhm water is filtered using a 0.2 micron filter, and filled into sterile bottles.


  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
  • High Quality - No detectable DNase, RNase, or protease activities.
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs


Water is a key component in so many experiments and pure, quality controlled and traceable water can help you improve your control over critical, sensitive experiments. Lonza’s AccugeneTM Molecular Biology Grade water has been the reagent of choice for decades for electrophoresis, cloning, PCR and other applications that demand the strictest of quality controls.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR