AccuGENE™ Buffers
Supplier: LONZA WALKERSVILLE INC. CA
51205
51214
CA12001-388EA
87.9
CAD
CA12001-388
CA12001-394
AccuGENE™ Buffers
Buffers
Electrophoresis Buffers
Hybridisation Buffers
AccuGENE® buffers are prepared with high quality reagents and use 18 megohm water
- Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
- High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
- Efficient - Ready-made solutions eliminate experiment preparation time
- Flexible - Customized solutions are available to meet individual needs
- Formulation - 3.0 M NaCl, 0.3 M sodium citrate, pH 7.0
Store all reagents at 18 to 26 °C (64 to 79 °F).
They are manufactured according to strict quality control standards to ensure lot-to-lot consistency.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.