Pierce BupH™ Dry Blend Buffers, Washing, Thermo Scientific
Supplier: Thermo Scientific
28376 28372 28374 28379 28382 28384 28390
CAPI-28376PK 392.32 CAD
CAPI-28376 CAPI28372 CAPI28374 CAPI28379 CAPI28382 CAPI28384 CAPI28390
Pierce BupH™ Dry Blend Buffers, Washing, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers
BupH™ packs are pre-blended and pre-measured dry mixtures of buffers that are easy to prepare and used in protein purification and common laboratory techniques.
  • Packs are pouches of dry-blend powder that are each sufficient to make 500 mL of buffer
  • Eliminates weighing time and pH adjustments
  • Every pack will yield the same, consistent buffer
  • Eliminates concerns about long-term stability of stock solutions