VWR® Contour™ Standing Height Base Cabinet for Acid Storage
Supplier: VWR International
CFG-2804-A CFG-2804-L CFG-2805-A CFG-2806-A CFG-2808-A CWG-2804-A CWG-2804-L CWG-2805-A CWG-2806-A CWG-2808-A
CFG-2804-AEA 4142.51 CAD
CFG-2804-A CFG-2804-L CFG-2805-A CFG-2806-A CFG-2808-A CWG-2804-A CWG-2804-L CWG-2805-A CWG-2806-A CWG-2808-A
VWR® Contour™ Standing Height Base Cabinet for Acid Storage
Furniture Casework
Heavy-gauge steel full-frame construction, specifically designed for the storage of corrosive chemicals.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
  • Lined with a molded one piece linear LDPE tub with coved corners and a 1" lip
  • Cabinet doors are lined with 18" PE sheet
  • Doors are latched using a nylon roller catch
Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above