Sklar Edge™ Tungsten Carbide Mayo-Stille Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
16-3552 16-3557 16-3562 16-3567
89025-732EA 603.61 CAD
89025-732 89025-734 89025-736 89025-738
Sklar Edge™ Tungsten Carbide Mayo-Stille Dissecting Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Mayo-Stille Dissecting Scissors feature tungsten carbide inserts that are permanently bonded to the scissor blade
This provides an exceptionally hard and durable cutting edge and increases the overall life the instrument. Mayo-Stille Dissecting Scissors have blunt tips and are available curved or straight. The edges are rounded which permits deeper dissection of tough tissue.