Rubber Stopper for Serum Bottles, Ace Glass Incorporated
Supplier: Ace Glass
5531-06 5531-23 5531-33
89051-750CS 1257.32 CAD
89051-750 89051-752 89051-754
Rubber Stopper for Serum Bottles, Ace Glass Incorporated
Stoppers Rubber Stoppers
Rubber stoppers of various styles and rubber compounds for serum vials and bottles
Stoppers fit a mouth I.D. of 13mm and an O.D. of 20mm. All stoppers fit 20mm bottle and vial flanges.