Mayo Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
22-1556 22-1567 22-2567
89025-268EA 225.65 CAD
89025-268 89025-270 89025-290
Mayo Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklar® Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings
They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences.
Order Now
SPECIFICATIONS
Environmentally Preferable