TC Mayo-Hegar Left Handed Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-9016 21-9018
10195-064EA 687.49 CAD
10195-064 10195-066
TC Mayo-Hegar Left Handed Needle Holder, OR Grade, Sklar
Forceps
The Sklar® Left-Handed TC Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures
Due to it's versatility, the Mayo-Hegar needle holder it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Tungsten carbide inserts also maximize control and precision. This product is straight, with cross-serrated jaws, and an overall length of 6 inches. It features a unique ratchet design for left handed use.