Biopsy Nylon Mesh Bags, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
BIO3045W BB0023
10809-874PK 252.33 CAD
10809-874 10809-876 10809-890
Biopsy Nylon Mesh Bags, Cancer Diagnostics
Bags Sample Bags
Biopsy nylon mesh bags use special material for effortless opening during embedding.
  • Folded bottom
  • Gentle seams
  • Twist open bag effortlessly
Material makes it effortless to twist open bag between thumb and forefinger.
Blue biopsy bags are colored blue to help facilitate location of small biopsy specimens following processing and are available in dispenser packs for convenience.
Biopsy cassette pads use thin, non-stick foam (1×1.125") to allow for optimal fluid transfer with virtually no carryover. Pressure on biopsy is greatly reduced decreasing occurrence of artifact.
Folded bottom and gentle seams make for easy opening during embedding greatly reducing the 'springboard effect' associated with other bags.