Blueing reagent working solution, Pureview PH-Blue (pH 8)
Supplier: CANCER DIAGNOSTICS, INC.
16702-4
10809-908CS
739.95
CAD
10809-908
Blueing reagent working solution, Pureview PH-Blue (pH 8)
Stains
Microscopy and staining reagents
The Endure Staining System is designed to produce precise, consistent H&E staining.
A bluing reagent buffered to a constant pH 8. Fluctuations in pH have been associated with “floaters” and unclear nuclear detail. PH Blue is designed to be unaffected by carryover and is formulated to help standardize the staining process. Available in working solution or concentrate formula.