Vapor Monitor Badges, Cancer Diagnostics
Supplier: Cancer Diagnostics
XVM-2 FVM-2 XVM-6 FVM-6
10809-888CS 2085.77 CAD
10809-888 10809-886 10809-884 10809-882
Vapor Monitor Badges, Cancer Diagnostics
Gas Detection Systems
Vapor Monitor Badges easily monitor your breath and air quality.
  • Foil seal
  • Zip lock pouch
  • Badges include report
Monitor your air quality and breath easy! Badges include report. Take badge out of foil seal and zip lock pouch and clip on collar/shirt. Wear for 15 minutes or 8 hours(short term/long term exposure) and place back in pouch. Send to CDI's independent lab in pre-addressed envelope and receive a report in 48 hours.