Tissue Marking Dye Kit, Cancer Diagnostics
Supplier: Cancer Diagnostics
04000 03000P 06000 03000
10809-868KT 902.7 CAD
10809-868 10809-866 10809-872 10809-870
Tissue Marking Dye Kit, Cancer Diagnostics
Stains Histology Stains
The Original 7 Dye Color Kit is used for defining tissue margins and multiple biopsies.
  • Defines tissue margins
  • Consistent
  • Durable
  • Diagnostic assurance
Tissue marking dyes for marking margins & multiple biopsies. The Original 7 Dye Color Kit for defining tissue margins. Consistency, durability, and brightness under the microscope provide diagnosing assurance. A variety of packaging options provide greater application options and efficiency.
Packaging: Pack of 7