Tissue Marking Dye, Flip-Top, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
0723-1 0723-2 0723-3 0723-4 0728-6 0728-7 0727-2 0728-1 0728-2 0728-3 0728-5 0727-3 0727-1 0727-4
10809-812EA 167.91 CAD
10809-812 10809-814 10809-816 10809-818 10809-820 10809-822 10809-824 10809-826 10809-828 10809-830 10809-846 10809-848 10809-850 10809-852 10809-854 10809-856 10809-858 10809-860 10809-862 10809-864 10809-912 10809-914 10809-916 10809-918 10809-920 10809-922 10809-924 10809-926 10810-000 10810-002
Tissue Marking Dye, Flip-Top, Cancer Diagnostics
Stains Histology Stains
Tissue marking dyes are used for marking margins & multiple biopsies.
  • Margins defined without bleeding
  • Dyes won't change color or fade
  • Flip-top dispenser cap
Tissue marking dyes for marking margins & multiple biopsies. Dyes have been developed to define margins without bleeding, changing color or fading. Flip-top dispenser cap.