Nesting Container with Hand Holes, MFG Tray
Supplier: MFG Tray
912118-5269 912148-5269 912248-5269
89022-914EA 304.39 CAD
89022-914 89022-918 89022-916
Nesting Container with Hand Holes, MFG Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
MFG Tray's durable nesting containers are resistant to cutting oils, grease, and solvents
Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for work-in-process part handling. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats, and totes for material handling in the metalworking, plastics, and electronics assembly industries, as well as in food service, confectionery, and pharmaceutical processing. MFG Tray products combine outstanding performance features with long service life to ensure exceptional value. The material matters in material handling.