ESD Stacking Box, MFG
Supplier: MFG Tray
8480005167
89022-942EA 202.08 CAD
89022-942
ESD Stacking Box, MFG
Boxes ESD-Safe Boxes
MFG Tray ESD stacking boxes provide optimal product protection with exceptional weight-carrying capability
Boxes possess superior physical properties over thermoplastic and corrugated products. They will not bend or sag under heavy loads and can be readily integrated into automated applications. ESD stacking boxes ensure optimum space utilization during assembly operations, and may be used to form mobile work stations. Fiberstat stacking boxes ensure complete protection from electrostatic discharge for the life of the product. They are resistant to cutting oils, greases, solvents, mild acids, and alkaline solutions within a pH rating of 3.0 to 10.0. Stacking boxes are easily cleaned in hot water or steam with standard detergents. Lids are available separately for certain models.