Toteline Nest and Stack Container Dolly, MFG
Supplier: Molded Fiber Glass Tray
780138-5172 780238-5172 780338-5172 780438-5172
89022-848EA 601.06 CAD
89022-848 89022-854 89022-860 89022-866
Toteline Nest and Stack Container Dolly, MFG
Trolleys
Heavy-duty fiberglass dollies will easily transport the contents of MFG Tray's durable Stacking containers
These dollies and containers are resistant to cutting oils, grease, and solvents. Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for conveying heavy stacks of work-in-process parts on the standard 3" casters. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable