Conveyor/Assembly Tray, MFG Tray
Supplier: Molded Fiber Glass Tray
3320011311 6290085136
89177-116EA 122.91 CAD
89177-116 89177-120
Conveyor/Assembly Tray, MFG Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
MFG Tray conveyor and assembly trays feature a low-profile design and radial edges to readily accommodate conveyor transport and assembly operations in a wide variety of applications
Trays are designed for component assembly and for compatability with conveyors or cart transport systems.
Four distinct corner profiles are available to integrate with rack systems and conveyors, or for nesting or stacking capabilities. Their dimensional stability and light weight ensure easy transporting and handling. Composite construction enables these trays to be constantly used in temperatures rangingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
pH range 3.0 to 10.0
Temperature range –51 to 121°C (–60 to 250°F), up to 300°F intermittently