InLab® Sensors and Cables, METTLER TOLEDO
Supplier: Mettler Toledo
30281896 30281897 30281898 30281899 30281910 30281911 30281913 30281915 30281916 30281917 30281918 30281919 30281920 30281921 30281922 30281923 30281924 30281925 30281926 30281927 30281928 30248830 30248832 30244733 30248120 30248826 30248828 30244732 30248112 30247850 30249570 30253098
CA10842-580EA 264.81 CAD
CA10842-580 CA10842-582 CA10842-592 CA10842-594 CA10842-596 CA10842-598 CA10842-600 CA10842-602 CA10842-604 CA10842-606 CA10842-608 CA10842-610 CA10842-612 CA10842-614 CA10842-616 CA10842-618 CA10842-620 CA10842-622 CA10842-624 CA10842-626 CA10842-628 CA10842-630 CA10842-632 CA10842-634 CA10842-636 CA10842-638 CA10842-640 CA10842-642 CA10842-646 CA10842-648 CA10842-650 CA10842-652 CA10842-654
InLab® Sensors and Cables, METTLER TOLEDO
Electrodes pH Electrodes
METTLER TOLEDO electrodes combine innovative technologies for fast, accurate and reliable measurements.
  • Easy to use
  • Built to last
  • Performance to trust
All sensors are shipped ready to use
The well-designed InLab sensor construction, as well as the high-quality materials used for manufacturing guarantee a robust sensor - even in harsh applications. The versatile sensor portfolio ensures the prefectly suited sensor for every application, a match that will greatly lengthen the lifetime of the product.
Quality is more than a promise for us. To guarantee it, we test every single sensor. Only sensors which successfully pass the strict final product inspection receive an individual quality certificate, and are ready for delivery.
Everything you need to measure comes together with the sensor. Your work in the lab will be simplified by the spill-proof wetting cap, the automatic sensor recognition of ISM sensors, and the application-specificity of the sensor you select.