ESD Conveyor/Assembly Tray, MFG Tray
Supplier: Molded Fiber Glass Tray
3030005167 3180005167 2280005167 3320005167
89022-930EA 80.06 CAD
89022-930 89022-934 89022-944 89177-118
ESD Conveyor/Assembly Tray, MFG Tray
Trays/Dishes/Pots Laboratory Trays/Dishes
Used in the assembly of circuit boards, computers, medical device, and other types of electronic or static sensitive assembly
Trays are designed for component assembly and for compatability with conveyors or cart transport systems.
Order Now
SPECIFICATIONS
Environmentally Preferable
pH range 3.0 to 10.0
Temperature range –51 to 121°C (–60 to 250°F), up to 300°F intermittently