Volkman Bone Curette, OR Grade, Sklar®
Supplier: Sklar
40-7250 40-7265 40-7280
10196-744EA 353.46 CAD
10196-744 10196-746 10196-764
Volkman Bone Curette, OR Grade, Sklar®
Curettes
Sklar's® Volkman Bone Curette is a debulking instrument commonly used in orthopedic procedures.
It is used to scrape away and remove fibrous tissue and other debris from bone. It may also be used to scrape and remove small portions of the bone itself.
Ordering information: Volkmann bone curettes are available as single or double-ended instruments, in various lengths, with multiple tip configurations.