VWR® Contour™ Wall Mounted Storage Cabinets, Pass-Thru Storage Cases
Supplier: VWR International
CFW13301336-A CFW13301348-A CWW13301336-A CWW13301348-A
CFW13301336-AEA 3716.98 CAD
CFW13301336-A CFW13301348-A CWW13301336-A CWW13301348-A
VWR® Contour™ Wall Mounted Storage Cabinets, Pass-Thru Storage Cases
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Wall cabinets come with two adjustable shelves.
Ordering information: Installation is required.
Order Now
SPECIFICATIONS
Environmentally Preferable