Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Stainless Steel, Metro
Supplier: Metropolitan Wire
A1824NS A1830NS A1836NS A1842NS A1848NS A1854NS A1860NS A1872NS A2424NS A2430NS A2436NS A2442NS A2448NS A2454NS A2460NS A2472NS A1424NS A1430NS A1436NS A1442NS A1448NS A1460NS A1472NS A2124NS A2130NS A2136NS A2142NS A2148NS A2154NS A2160NS A2172NS A1424NS A1430NS A1436NS A1442NS A1448NS A1460NS A1472NS A2124NS A2130NS A2136NS A2142NS A2148NS A2154NS A2160NS A2172NS
CA19766-940EA 874.44 CAD
CA19766-940 CA19766-942 CA19766-944 CA19766-946 CA19766-948 CA19766-950 CA19766-952 CA19766-954 CA19766-956 CA19766-958 CA19766-960 CA19766-962 CA19766-964 CA19766-966 CA19766-968 CA19766-970 89369-758 89369-764 89369-770 89369-776 89369-782 89369-788 89369-794 89369-800 89369-806 89369-812 89369-818 89369-824 89369-830 89369-836 89369-842 CA89369-758 CA89369-764 CA89369-770 CA89369-776 CA89369-782 CA89369-788 CA89369-794 CA89369-800 CA89369-806 CA89369-812 CA89369-818 CA89369-824 CA89369-830 CA89369-836 CA89369-842
Super Adjustable 2™ Super Erecta Shelf® Wire Shelving, Stainless Steel, Metro
Furniture Shelves
Type 304 stainless steel shelves provide maximum protection in corrosive environments.
  • Adjustable shelves with quick and easy shelf adjustment
  • Design own wire shelf system with appropriate accessories
Notes: The actual length of the shelves is 3.2 mm (¹/₈") shorter than the nominal dimension shown. Stationary posts over 189.6 cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. The 245.4 cm high stationary posts areMore Product Information