Super Erecta Shelf® Ledges, 100 mm (4"), Stackable, Metro™
Supplier: Metropolitan Wire
L18N-4C L21N-4C L24N-4C L30N-4C L36N-4C L42N-4C L48N-4C L54N-4C L60N-4C L72N-4C L18N-4S L21N-4S L24N-4S L30N-4S L36N-4S L42N-4S L48N-4S L54N-4S L60N-4S L72N-4S L14N-4C L14N-4S
CA22233-274EA 70.52 CAD
CA22233-274 CA22233-276 CA22233-278 CA22233-280 CA22233-282 CA22233-284 CA22233-286 CA22233-288 CA22233-290 CA22233-292 CA22233-296 CA22233-298 CA22233-300 CA22233-302 CA22233-304 CA22233-306 CA22233-308 CA22233-310 CA22233-312 CA22233-314 CA22233-272 CA22233-294
Super Erecta Shelf® Ledges, 100 mm (4"), Stackable, Metro™
Furniture Shelves
Stackable Super Erecta Shelf® Ledges for use with Super Erecta® shelving systems.
  • May be stacked to form an 8" (203 mm) or higher barrier
  • Ledges prevent items from protruding or falling from shelves
Note: Actual ledge length is approximately 2.5 cm (1") shorter than the nominal shelf length/width.