Key Elevator, OR Grade, Sklar®
Supplier: Sklar
40-7402 40-7398 40-7400 40-7401 40-7403 40-7404 40-7406
10194-068EA 311.58 CAD
10194-068 10196-842 10196-844 10196-846 10196-848 10196-850 10196-884
Key Elevator, OR Grade, Sklar®
Retractors
Sklar's® Key Elevator is used in neurological procedures for lifting soft tissue and periosteum.
  • Premium OR-Grade
It can also be used to lift bone in order to clear the surgery site. It features a solid, hexagonal handle with a gently curved blade tip with a sharp edge. It is available in several lengths with varying blade widths.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.