Fosfomycin disodium salt 90%

Supplier: Thermo Scientific Chemicals

J66602-03 J66602-06
AAJ66602-03EA 95.61 CAD
AAJ66602-03 AAJ66602-06
Fosfomycin disodium salt 90%
Fosfomycin disodium salt

Fosfomycin disodium prevents peptidoglycan synthesis by inhibiting MurA, an enzyme responsible for synthesizing N-acetylmuramic acid or NAM, a major component of peptidoglycan.


  • White to off-white powder


White to off-white powder


Freely soluble in water or aqueous solution

Formula: C₃H₅Na₂O₄P
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056853
CAS Number: 26016-99-9
EINECS: 247-409-2

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR