Aloin
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one, Barbaloin, 1,8-Dihydroxy-10-(beta-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone, Aloin A, 10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone
Aloe exudant, used as a stimulant-laxative
CAS: 1415-73-2
Form: Powder
Color: Yellow to brown
MDL Number: MFCD00151160
Molecular Formula: C21H22O9
Formula weight: 418.40
Synonyms: Barbaloin, 1,8-Dihydroxy-10-(beta-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone
Savety Data Phrase: Warning
Beilstein Reference: 6077558
GHS H Statement:H315-H319-H335
GHS P Statement: P261-P280-P305+P351+P338-P304+P340-P405-P501a
Formula:
C₂₁H₂₂O₉ Storage Temperature: Ambient |
MDL Number:
MFCD00151160 CAS Number: 1415-73-2 EINECS: 215-808-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...