Weitlaner-Beckman Retractor, OR Grade, Sklar®
Supplier: Sklar
42-1655 42-1765
10196-686EA 1533.47 CAD
10196-686 10196-688
Weitlaner-Beckman Retractor, OR Grade, Sklar®
Retractors
Sklar's® Weitlaner-Beckmann Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site
It has curved shanks with hinged arms that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed. The hinged arms of the Weitlaner-Beckmann retractor allow the blades to be used in difficult to reach surgical sites, such as in spinal or intracranial procedures.