Carbon Film on Thin Bar Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFT300-CU-25 CFT300-CU-UL CFT400-CU-UL CFT1000-CU-UL CFT300-CU-TH CFT400-CU-TH CFT1000-CU-TH CFT300-CU-ET CFT400-CU-ET CFT1000-CU-ET CFT200-NI-UL CFT300-NI-UL CFT400-NI-UL CFT1000-NI-UL CFT200-CU-TH CFT200-CU-ET CFT400-NI-TH CFT1000-NI-TH CFT200-AU-UL CFT300-AU-UL CFT400-AU-UL CFT200-CU-UL CFT200-NI-TH CFT300-NI-TH CFT200-NI-ET CFT300-NI-ET CFT400-NI-ET CFT1000-NI-ET CFT1000-AU-UL CFT1000-AU-TH CFT200-AU-ET CFT300-AU-ET CFT400-AU-ET CFT1000-AU-ET CFT200-AU-TH CFT300-AU-TH CFT400-AU-TH
102100-314BX 239.4 CAD
102100-314 103304-826 103304-828 103304-830 103304-904 103304-906 103304-908 103304-976 103304-978 103304-980 103305-512 103305-514 103305-516 103305-518 103305-548 103305-550 103305-594 103305-596 103305-874 103305-876 103305-878 103305-976 103306-020 103306-022 103306-068 103306-070 103306-072 103306-074 103306-110 103306-124 103306-128 103306-130 103306-132 103306-358 103306-366 103306-368 103306-370
Carbon Film on Thin Bar Square Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Select copper, nickel, or gold grids in 200, 300, 4000, or 1000 mesh
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 - 6 nm), thick (15 - 20 nm), or extra thick (25 - 50 nm).
Packaging: Packed in grid storage box.