Carbon Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFTH400-CU-50 CFTH600-CU-25 CFTH300-CU-UL CFTH400-CU-UL CFTH600-CU-UL CFTH300-CU-TH CFTH400-CU-TH CFTH600-CU-TH CFTH300-CU-ET CFTH400-CU-ET CFTH600-CU-ET CFTH200-NI-UL CFTH300-NI-UL CFTH400-NI-UL CFTH600-NI-UL CFTH200-CU-TH CFTH200-CU-ET CFTH200-NI-TH CFTH300-NI-TH CFTH200-CU-UL CFTH400-NI-TH CFTH600-NI-TH CFTH200-NI-ET CFTH300-NI-ET CFTH400-NI-ET CFTH600-NI-ET CFTH200-AU-UL CFTH300-AU-UL CFTH300-AU-TH CFTH400-AU-UL CFTH600-AU-UL CFTH300-AU-ET CFTH400-AU-TH CFTH600-AU-TH CFTH200-AU-ET CFTH400-AU-ET CFTH600-AU-ET CFTH200-AU-TH
102100-364BX 273.07 CAD
102100-364 102100-374 103304-832 103304-834 103304-836 103304-910 103304-912 103304-914 103304-982 103304-984 103304-986 103305-520 103305-522 103305-524 103305-526 103305-552 103305-554 103305-598 103305-600 103305-978 103306-024 103306-026 103306-076 103306-078 103306-080 103306-082 103306-112 103306-114 103306-116 103306-118 103306-120 103306-134 103306-136 103306-138 103306-360 103306-362 103306-364 103306-372
Carbon Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Select copper, nickel, or gold grids in 200, 300, 400, or 600 mesh
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.