Carbon Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CF400-NI-50 CF200-NI-50 CF300-NI-50 CF400-CU-25 CF200-AU-50 CF300-AU-25 CF200-CU-50 CF400-CU-50 CF150-CU-50 CF300-AU-50 CF400-AU-25 CF300-CU-25 CF300-CU-50 CF200-CU-25 CF150-AU-50 CF400-AU-50 CF150-CU-25 CF200-CU-TH CF300-CU-TH CF400-CU-TH CF200-CU-ET CF400-CU-ET CF150-CU-UL CF200-CU-UL CF300-CU-UL CF400-CU-UL CF200-AU-UL CF400-AU-UL CF400-AU-TH
100500-858BX 292 CAD
100500-858 100489-726 100500-848 100503-154 100489-720 100489-728 100489-722 100500-854 100489-712 100489-730 100500-850 100502-760 100489-732 100502-758 100489-710 100500-852 100502-756 103303-420 103303-422 103303-424 103303-498 103303-502 103303-534 103303-536 103303-538 103303-540 103305-962 103305-966 103305-998
Carbon Film on Square Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Select copper, nickel, or gold grids in 150, 200, 300, or 400 mesh
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of Standard: Approx. 5-6nm, Ultra-Thin (UL): 3-4nm, Thick (TH): 10nm, Extra Thick (ET): 20-30nm.
Ordering information: Available in Standard (5 - 6 nm), Ultra-Thin (3 - 4 nm), Thick (10 nm) and Extra Thick (20 - 30 nm) film thickness.
Packaging: Packed in grid storage box.
Order Now
SPECIFICATIONS
Environmentally Preferable