Carbon Film on Slots, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CF205-CU-UL CF2010-CU-UL CF205-NI-UL CF205-CU-TH CF2010-CU-TH CF205-CU-ET CF2010-CU-ET CF2010-NI-UL CF205-NI-TH CF2010-NI-TH CF205-NI-ET CF2010-NI-ET CF205-AU-UL CF2010-AU-UL CF205-AU-TH CF2010-AU-TH CF205-AU-ET CF2010-AU-ET
103304-016EA 320.11 CAD
103304-016 103304-018 103304-238 103304-390 103304-392 103304-474 103304-476 103304-774 103304-838 103304-840 103304-916 103304-918 103305-224 103305-226 103305-364 103305-366 103305-446 103305-448
Carbon Film on Slots, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Optically checked followed by batch testing in the EM
Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.