Carbon Film on Single Hole Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFGA75-CU-UL CFGA100-CU-UL CFGA150-CU-UL CFGA200-CU-UL CFGA300-CU-UL CFGA400-CU-UL CFGA600-CU-UL CFGA800-CU-UL CFGA1000-CU-UL CFGA1500-CU-UL CFGA75-CU-TH CFGA100-CU-TH CFGA150-CU-TH CFGA200-CU-TH CFGA300-CU-TH CFGA400-CU-TH CFGA600-CU-TH CFGA800-CU-TH CFGA1000-CU-TH CFGA1500-CU-TH CFGA75-CU-ET CFGA100-CU-ET CFGA150-CU-ET CFGA200-CU-ET CFGA600-CU-ET CFGA1000-CU-ET CFGA1500-CU-ET CFGA75-NI-UL CFGA100-NI-UL CFGA150-NI-UL CFGA200-NI-UL CFGA300-CU-ET CFGA300-NI-UL CFGA400-CU-ET CFGA400-NI-UL CFGA600-NI-UL CFGA800-CU-ET CFGA800-NI-UL CFGA1000-NI-UL CFGA1500-NI-UL CFGA75-NI-TH CFGA100-NI-TH CFGA150-NI-TH CFGA200-NI-TH CFGA300-NI-TH CFGA400-NI-TH CFGA600-NI-TH CFGA800-NI-TH CFGA1000-NI-TH CFGA1500-NI-TH CFGA75-NI-ET CFGA100-NI-ET CFGA150-NI-ET CFGA200-NI-ET CFGA300-NI-ET CFGA400-NI-ET CFGA600-NI-ET CFGA800-NI-ET CFGA1000-NI-ET CFGA1500-NI-ET
103304-020EA 308.35 CAD
103304-020 103304-022 103304-024 103304-026 103304-028 103304-030 103304-032 103304-034 103304-036 103304-038 103304-394 103304-396 103304-398 103304-400 103304-402 103304-404 103304-406 103304-408 103304-410 103304-412 103304-478 103304-480 103304-482 103304-484 103304-486 103304-488 103304-490 103304-776 103304-778 103304-780 103304-782 103304-784 103304-786 103304-788 103304-790 103304-792 103304-794 103304-796 103304-798 103304-800 103304-842 103304-844 103304-846 103304-848 103304-850 103304-852 103304-854 103304-856 103304-858 103304-860 103304-920 103304-922 103304-924 103304-926 103304-928 103304-930 103304-932 103304-934 103304-936 103304-938
Carbon Film on Single Hole Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Carbon coatings thickness of standard (10 nm), ultra-thin (5 – 6 nm), thick (15 – 20 nm), or extra thick (25 – 50 nm)
  • Select from copper and nickel with 75, 100, 150, 200, 300, 400, 600, 800, 1000, or 1500 µm holes
Optically checked followed by batch testing in the electron microscope.
Packaging: Packed in grid storage box.