Merit™ Graves Vaginal Speculum, Physician Grade, Sklar®
Supplier: Sklar
98-303 98-312 98-324 98-325
10185-298EA 59.37 CAD
10185-298 10193-774 10193-976 75996-194
Merit™ Graves Vaginal Speculum, Physician Grade, Sklar®
Specula
The Merit™ Graves vaginal speculum is a gynecological instrument used for performing pelvic examinations, Pap smears, and examining the cervix.
  • Durable
  • Wider blades than Pederson speculum
  • Cost effective and high quality
This is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient.
Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures. These are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments.