DeBakey Bulldog Clamp, OR Grade, Sklar®
Supplier: Sklar
52-1530 52-1531 52-1533 52-1540 52-1541 52-1542 52-1543
89025-130EA 537.54 CAD
89025-130 89025-132 89025-136 89025-138 89025-140 89025-142 89025-144
DeBakey Bulldog Clamp, OR Grade, Sklar®
Forceps
Sklar's® DeBakey Atraumatic Bulldog Clamp is a small cross-action thumb clamp used for halting blood flow in a vessel
They are most commonly used in cardiovascular or cardiothoracic procedures. They are available curved or straight, in multiple lengths