Hoff Towel Clamp, OR Grade, Sklar®
Supplier: Sklar
47-3040 47-3057
89025-042EA 313.53 CAD
89025-042 89025-044
Hoff Towel Clamp, OR Grade, Sklar®
Forceps
Sklar® Hoff Towel Clamps are a non-perforating clamp used to secure instruments and towels to the drape surrounding a surgical site
The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The tip of this towel clamp are round with a cross-serrated surface so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. This clamp should never be used on skin.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.