Plastic Surgery Retractor, OR Grade, Sklar®
Supplier: Sklar
22-9132 22-9133 22-9142 22-9143
89025-430EA 262.75 CAD
89025-430 89025-432 89025-434 89025-436
Plastic Surgery Retractor, OR Grade, Sklar®
Retractors
Sklar's® Plastic Surgery Retractor is a handheld retractor used in plastic surgery procedures for retracting tissue and exposing the surgical site
It has a straight handle, flexible shank, and is available with two or three sharp or blunt prongs.