Miller-Senn Retractor, OR Grade, Sklar®
Supplier: Sklar
22-8360 22-8460
89025-418EA 199.71 CAD
89025-418 89025-420
Miller-Senn Retractor, OR Grade, Sklar®
Retractors
Sklar's® Miller-Senn Double-Ended Retractor, which may also simply be known as a Senn retractor, is a handheld retractor with one L-shaped Moberg blade and one three pronged end that face opposite directions.
The solid blade end measures approximately 5 x 18 mm and the prong end measures 7 x 18 mm. The prongs are available sharp or blunt. This retractor is often used for holding open tissue or muscle in small surgical sites.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.