Deaver Retractor, OR Grade, Sklar
Supplier: Sklar
22-8080 22-8180 60-3012 60-2710 60-2712 60-2713 60-2812 60-2912 60-3070 60-3085 60-3090 60-3112 60-3113 60-3114 60-3212 60-3312 60-3412
89025-400EA 295.32 CAD
89025-400 89025-402 10194-034 10197-306 10197-308 10197-310 10197-312 10197-314 10197-316 10197-318 10197-320 10197-722 10197-724 10197-726 10197-728 10197-730 10197-732
Deaver Retractor, OR Grade, Sklar
Retractors
The Deaver Retractor is a large, handheld retractor commonly used to hold back the abdominal wall during abdominal or thoracic procedures. It may also be used to move or hold organs away from the surgical site.
The Deaver retractor is shaped like a question mark with a thin, flat blade
Censitrac Ready products are industry standard and constructed of stainless steel.
It is available with a grip handle which is often held by a surgical assistant during procedures, or a flat handle so the retractor can be clamped into a position.
Ordering information: Censitrac instruments areMore Product Information